Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195286.1 | NP_001182215.1 | 5, 6 | 769 | ||
NM_002460.3 | NP_002451.2 | 5, 6 | 772 | |||
NR_046000.2 | 5, 6 | 769 | ||||
GenBank mRNA | AK130762.1 | - | 2, 3 | 663 | ||
AK310149.1 | - | 5, 6 | 759 | |||
BC015752.1 | - | 5, 6 | 719 | 4861223 | ||
D78261.1 | - | 5, 6 | 738 | |||
U52682.1 | - | 5, 6 | 768 | |||
U63738.1 | - | 5, 6 | 751 |