Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002203.3 | NP_002194.2 | 5 | 557 | ||
NR_073103.1 | 5 | 557 | ||||
NR_073104.1 | 5 | 557 | ||||
NR_073105.1 | 5 | 557 | ||||
NR_073106.1 | 5 | 557 | ||||
NR_073107.1 | 4 | 436 | ||||
GenBank mRNA | AK307952.1 | - | 5 | 546 | ||
BC143505.1 | - | 5 | 443 | |||
BC143508.1 | - | 5 | 443 | |||
BC143509.1 | - | 5 | 443 | |||
BC143511.1 | - | 5 | 443 | |||
BC143512.1 | - | 4 | 322 | |||
BE048389.1 | - | 5 | 562 | |||
BP280760.1 | - | 5 | 560 | |||
X17033.1 | - | 5 | 462 |