Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000885.5 | NP_000876.3 | 4, 5 | 1309 | ||
GenBank mRNA | AB208885.1 | - | 1, 2 | 88 | ||
AK303478.1 | - | 4, 5 | 796 | |||
BC016671.2 | - | 4, 5 | 774 | |||
L12002.1 | - | 4, 5 | 974 | |||
X16983.1 | - | 4, 5 | 588 |