Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002207.2 | NP_002198.2 | 23 | 2524 | ||
GenBank mRNA | D25303.1 | - | 24 | 2522 | ||
L24158.1 | - | 23 | 2366 |