Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002211.3 | NP_002202.2 | 9 | 1288 | ||
NM_033668.2 | NP_391988.1 | 8 | 1067 | |||
NM_133376.2 | NP_596867.1 | 9 | 1203 | |||
GenBank mRNA | AK291697.1 | - | 9 | 1140 | ||
BC020057.1 | - | 9 | 1132 | 3924411 | ||
BC113901.1 | - | 3 | 289 | |||
BX537407.1 | - | 9 | 1142 | |||
CX868374.1 | - | 5 | 545 | |||
CX871886.1 | - | 5 | 696 |