Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002214.2 | NP_002205.1 | 3 | 992 | ||
XM_011515393.2 | XP_011513695.1 | 4 | 795 | |||
XM_011515394.2 | XP_011513696.1 | 4 | 393 | |||
XM_011515396.1 | XP_011513698.1 | 4 | 1064 | |||
XM_017012178.1 | XP_016867667.1 | 4 | 689 | |||
XM_017012179.1 | XP_016867668.1 | 4 | 1148 | |||
XM_017012180.1 | XP_016867669.1 | 4 | 1441 | |||
XM_017012181.1 | XP_016867670.1 | 5 | 1260 | |||
XM_017012182.1 | XP_016867671.1 | 4 | 375 | |||
XM_017012183.1 | XP_016867672.1 | 4 | 332 | |||
GenBank mRNA | AB209429.1 | - | 1 | 2440 | ||
AK295044.1 | - | 4 | 1064 | |||
BC002630.2 | - | 3 | 992 | 3609490 | ||
BI861225.1 | - | 3 | 626 | |||
BT006739.1 | - | 3 | 287 | |||
M73780.1 | - | 3 | 967 |