Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021262.1 | XP_016876751.1 | 3, 4 | 563 | ||
GenBank mRNA | AF279372.1 | - | 4, 5 | 427 | ||
AK293515.1 | - | 4, 5 | 482 | |||
BC003622.2 | - | 5, 6 | 587 | 3010239 | ||
BC007428.2 | - | 5, 6 | 587 | 3010239 | ||
BC018192.1 | - | 5, 6 | 536 | 3867269 | ||
BC037305.2 | - | 3, 4 | 453 | |||
BX407816.2 | - | 5, 6 | 652 | |||
BX420122.2 | - | 5, 6 | 651 | |||
CB997740.1 | - | 505 | ||||
U51336.1 | - | 5, 6 | 480 |