Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320923.1 | NP_001307852.1 | 23 | 3197 | ||
NM_001321852.1 | NP_001308781.1 | 22 | 3200 | |||
NM_001321853.1 | NP_001308782.1 | 24 | 3459 | |||
NM_001321854.1 | NP_001308783.1 | 23 | 3375 | |||
NM_001321855.1 | NP_001308784.1 | 23 | 3378 | |||
NM_001321856.1 | NP_001308785.1 | 22 | 3113 | |||
NM_001321857.1 | NP_001308786.1 | 22 | 3291 | |||
NM_002227.3 | NP_002218.2 | 22 | 3294 | |||
XM_017001212.1 | XP_016856701.1 | 22 | 3138 | |||
GenBank mRNA | AB209057.1 | - | 21 | 3050 | ||
AB219242.1 | - | 20 | 2941 | |||
AK314257.1 | - | 23 | 3198 | |||
BC132729.1 | - | 21 | 3012 | 40146751 | ||
BX647675.1 | - | 8 | 898 | |||
HQ628631.1 | - | 20 | 2974 | |||
M64174.1 | - | 21 | 3019 |