Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130089.1 | NP_001123561.1 | 12, 13 | 1649 | ||
NM_005548.2 | NP_005539.1 | 11, 12 | 1469 | |||
XM_017023217.1 | XP_016878706.1 | 11, 12 | 1561 | |||
GenBank mRNA | AF285758.1 | - | 12, 13 | 1627 | ||
AK126584.1 | - | 10, 11 | 925 | |||
AK315687.1 | - | 11, 12 | 1470 | |||
BC004132.2 | - | 11, 12 | 1432 | 2963032 | ||
BC013246.1 | - | 11, 12 | 1419 | |||
D31890.1 | - | 11, 12 | 1435 | |||
D32053.1 | - | 11, 12 | 1461 |