Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000217.2 | NP_000208.2 | 2 | 1686 | ||
GenBank mRNA | BC101733.1 | - | 1 | 619 | 8069239 | |
BC112180.1 | - | 1 | 619 | 8327648 | ||
L02750.1 | - | 1 | 581 |