Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002232.4 | NP_002223.3 | 1 | 1651 | ||
NR_109845.1 | 1 | 72 | ||||
GenBank mRNA | AK314009.1 | - | 1 | 1561 | ||
BC035059.1 | - | 1 | 1652 | 5261639 | ||
BC064595.1 | - | 1 | 1374 | |||
HY035404.1 | - | 1 | 73 | |||
L23499.1 | - | 1 | 1441 | |||
M85217.1 | - | 1 | 1354 |