Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002232.4 | NP_002223.3 | 1 | 1903 | ||
GenBank mRNA | AK314009.1 | - | 1 | 1813 | ||
BC035059.1 | - | 1 | 1904 | 5261639 | ||
BC064595.1 | - | 1 | 1626 | |||
L23499.1 | - | 1 | 1693 | |||
M85217.1 | - | 1 | 1606 |