Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004979.5 | NP_004970.3 | 1 | 1727 | ||
XM_011543910.2 | XP_011542212.1 | 2 | 635 | |||
XM_017029508.1 | XP_016884997.1 | 2 | 466 | |||
XM_017029509.1 | XP_016884998.1 | 2 | 463 | |||
XM_017029510.1 | XP_016884999.1 | 1 | 1577 | |||
GenBank mRNA | AB021865.1 | - | 1 | 1452 | ||
AF166003.1 | - | 1 | 204 | |||
AJ005898.1 | - | 1 | 154 | |||
AK315092.1 | - | 1 | 638 | |||
BC045659.1 | - | 1 | 1725 | 5271042 |