Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002238.3 | NP_002229.1 | 7, 8 | 1542 | ||
NM_172362.2 | NP_758872.1 | 7, 8 | 1623 | |||
XM_017001246.1 | XP_016856735.1 | 2, 3 | 761 | |||
GenBank mRNA | AF078741.1 | - | 7, 8 | 1408 | ||
AF078742.1 | - | 7, 8 | 1489 | |||
AJ001366.1 | - | 7, 8 | 1542 | |||
BC113709.1 | - | 7, 8 | 1442 | 8322761 | ||
BC143599.1 | - | 7, 8 | 1443 | 9052111 |