Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002241.4 | NP_002232.2 | 2 | 1191 | ||
GenBank mRNA | AK312383.1 | - | 2 | 1098 | ||
BC034036.1 | - | 2 | 1093 | |||
BC131627.1 | - | 2 | 1087 | 40118533 | ||
U52155.1 | - | 2 | 1177 | |||
U73193.1 | - | 1 | 526 |