Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002242.4 | NP_002233.2 | 2 | 350 | ||
GenBank mRNA | AF061118.1 | - | 2 | 301 | ||
AJ006128.1 | - | 1 | 256 | |||
AJ007557.1 | - | 2 | 329 | |||
AK314019.1 | - | 2 | 350 | |||
AY758241.1 | - | 1 | 232 | |||
BC037290.2 | - | 2 | 350 | 4817527 | ||
DA278958.1 | - | 2 | 348 |