Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172416.1 | NP_001165887.1 | 3 | 450 | ||
NM_001172417.1 | NP_001165888.1 | 3 | 453 | |||
NM_002242.4 | NP_002233.2 | 3 | 686 | |||
GenBank mRNA | AF061118.1 | - | 3 | 637 | ||
AJ006128.1 | - | 2 | 592 | |||
AJ007557.1 | - | 3 | 665 | |||
AK314019.1 | - | 3 | 686 | |||
AY758240.1 | - | 2 | 332 | |||
AY758241.1 | - | 2 | 568 | |||
BC037290.2 | - | 3 | 686 | 4817527 | ||
DB173405.1 | - | 3 | 450 | |||
DN993097.1 | - | 3 | 450 |