Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002245.3 | NP_002236.1 | 2 | 552 | ||
XM_011544184.2 | XP_011542486.1 | 4 | 718 | |||
GenBank mRNA | AK090902.1 | - | 5 | 751 | ||
AK314016.1 | - | 2 | 412 | |||
BC018051.1 | - | 2 | 429 | 4817185 | ||
U33632.1 | - | 2 | 566 | |||
U76996.1 | - | 2 | 413 | |||
U90065.1 | - | 2 | 566 |