Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004137.3 | NP_004128.1 | 3 | 597 | ||
GenBank mRNA | AF026002.1 | - | 2 | 154 | ||
AK313979.1 | - | 3 | 575 | |||
AY044441.1 | - | 3 | 220 | |||
AY515264.1 | - | 3 | 258 | |||
BC025707.1 | - | 3 | 554 | 5224514 | ||
U25138.1 | - | 3 | 313 | |||
U38907.1 | - | 2 | 154 | |||
U42600.1 | - | 3 | 207 | |||
U61536.1 | - | 3 | 259 |