Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004137.3 | NP_004128.1 | 4 | 766 | ||
GenBank mRNA | AF026002.1 | - | 3 | 323 | ||
AK313979.1 | - | 4 | 744 | |||
AY044441.1 | - | 4 | 389 | |||
AY515264.1 | - | 4 | 427 | |||
U25138.1 | - | 4 | 482 | |||
U38907.1 | - | 3 | 323 | |||
U42600.1 | - | 4 | 376 | |||
U61536.1 | - | 4 | 428 |