Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204824.1 | NP_001191753.1 | 10 | 1024 | ||
NM_004519.3 | NP_004510.1 | 10 | 1573 | |||
XM_006716555.3 | XP_006716618.1 | 8 | 834 | |||
XM_011517026.2 | XP_011515328.1 | 10 | 4107 | |||
XM_017013400.1 | XP_016868889.1 | 10 | 11079 | |||
GenBank mRNA | AB208890.1 | - | 10 | 1126 | ||
AF033347.1 | - | 10 | 1207 | |||
AK296293.1 | - | 10 | 1024 | |||
BC128576.1 | - | 11 | 1316 |