Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282428.1 | NP_001269357.1 | 3 | 1794 | ||
NM_002252.4 | NP_002243.3 | 3 | 1744 | |||
XM_011532825.1 | XP_011531127.1 | 4 | 2105 | |||
XM_017004059.1 | XP_016859548.1 | 3 | 1694 | |||
GenBank mRNA | AK314451.1 | - | 3 | 1744 |