Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002253.2 | NP_002244.1 | 30 | 4358 | ||
GenBank mRNA | AB209901.1 | - | 30 | 4343 | ||
AF035121.1 | - | 30 | 4359 | |||
AF063658.1 | - | 30 | 4056 | |||
AK293668.1 | - | 4 | 520 | |||
BC131822.1 | - | 30 | 4118 | 40129287 | ||
L04947.1 | - | 30 | 4050 | |||
X61656.1 | - | 30 | 4050 |