Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004521.2 | NP_004512.1 | 2 | 655 | ||
XM_017016224.1 | XP_016871713.1 | 2 | 261 | |||
GenBank mRNA | AK289413.1 | - | 2 | 601 | ||
AK307502.1 | - | 3 | 352 | |||
BC010885.1 | - | 2 | 591 | |||
BC065267.1 | - | 2 | 612 | |||
BC080604.1 | - | 2 | 586 | |||
BC111042.1 | - | 2 | 595 | |||
BC126279.1 | - | 2 | 408 | 8991995 | ||
BC126281.1 | - | 2 | 408 | 8991997 | ||
FJ224324.1 | - | 2 | 498 | |||
X65873.1 | - | 2 | 498 |