Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000222.2 | NP_000213.1 | 15 | 2260 | ||
NM_001093772.1 | NP_001087241.1 | 15 | 2248 | |||
XM_005265740.1 | XP_005265797.1 | 15 | 2234 | |||
XM_005265741.1 | XP_005265798.1 | 15 | 2231 | |||
XM_005265742.2 | XP_005265799.1 | 15 | 2222 | |||
XM_017008178.1 | XP_016863667.1 | 15 | 2228 | |||
XM_017008179.1 | XP_016863668.1 | 15 | 2219 | |||
XM_017008180.1 | XP_016863669.1 | 15 | 2216 | |||
GenBank mRNA | AJ438313.1 | - | 5 | 455 | ||
AK304031.1 | - | 15 | 2219 | |||
BC071593.1 | - | 15 | 2249 | 4375615 | ||
GU983671.1 | - | 15 | 2166 | |||
HM015525.1 | - | 15 | 2166 | |||
HM015526.1 | - | 15 | 2166 | |||
KT326922.1 | - | 15 | 2176 | |||
X06182.1 | - | 15 | 2194 |