Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256269.1 | NP_001243198.1 | 9 | 1255 | ||
NM_001256270.1 | NP_001243199.1 | 8 | 1343 | |||
NM_007317.2 | NP_015556.1 | 8 | 1206 | |||
GenBank mRNA | AB017430.2 | - | 8 | 1183 | ||
AK223431.1 | - | 8 | 1192 | |||
AK294380.1 | - | 8 | 1582 | |||
AK312234.1 | - | 8 | 1197 | |||
AK316050.1 | - | 8 | 1343 | |||
AK316389.1 | - | 9 | 1214 | |||
BC004352.1 | - | 8 | 1178 | 3535435 | ||
BC028155.1 | - | 8 | 1164 | 5241557 | ||
BT007259.1 | - | 8 | 1159 |