Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002268.4 | NP_002259.1 | 3 | 476 | ||
GenBank mRNA | AB002533.2 | - | 3 | 179 | ||
AK291041.1 | - | 3 | 428 | |||
BC028691.2 | - | 3 | 421 | 4838709 | ||
BC034493.2 | - | 3 | 476 | 4821602 | ||
U93240.1 | - | 3 | 320 |