Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021013.3 | NP_066293.2 | 2, 3 | 563 | ||
XM_011524793.1 | XP_011523095.1 | 3, 4 | 642 | |||
GenBank mRNA | BC041070.1 | - | 2, 3 | 582 | 5481838 |