Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002280.4 | NP_002271.3 | 7 | 1295 | ||
XM_011524794.1 | XP_011523096.1 | 7 | 2898 | |||
GenBank mRNA | X90763.1 | - | 7 | 1295 |