Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033033.3 | NP_149022.3 | 1 | 162 | ||
GenBank mRNA | AJ628419.1 | - | 1 | 125 |