Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000227.4 | NP_000218.3 | 12 | 1433 | ||
NM_001127717.2 | NP_001121189.2 | 48 | 6308 | |||
NM_001127718.2 | NP_001121190.2 | 11 | 1265 | |||
NM_198129.2 | NP_937762.2 | 49 | 6476 | |||
XM_011525978.2 | XP_011524280.1 | 49 | 6503 | |||
XM_011525979.2 | XP_011524281.1 | 49 | 6494 | |||
XM_011525980.2 | XP_011524282.1 | 49 | 6485 | |||
XM_011525981.2 | XP_011524283.1 | 48 | 6371 | |||
XM_011525982.2 | XP_011524284.1 | 49 | 6503 | |||
XM_017025743.1 | XP_016881232.1 | 32 | 4187 | |||
XM_017025744.1 | XP_016881233.1 | 22 | 2868 | |||
GenBank mRNA | AB107369.1 | - | 49 | 6235 | ||
AB208853.1 | - | 4 | 461 | |||
AY327114.1 | - | 11 | 1240 | |||
AY327115.1 | - | 49 | 6235 | |||
AY327116.1 | - | 48 | 6067 | |||
BC043618.1 | - | 6 | 813 | |||
L34155.1 | - | 12 | 1375 | |||
X84900.1 | - | 14 | 1654 | |||
X85107.1 | - | 12 | 1426 | |||
X85108.1 | - | 14 | 1654 |