Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000227.4 | NP_000218.3 | 17, 18 | 2211 | ||
NM_001127717.2 | NP_001121189.2 | 53, 54 | 7086 | |||
NM_001127718.2 | NP_001121190.2 | 16, 17 | 2043 | |||
NM_198129.2 | NP_937762.2 | 54, 55 | 7254 | |||
XM_011525978.2 | XP_011524280.1 | 54, 55 | 7281 | |||
XM_011525979.2 | XP_011524281.1 | 54, 55 | 7272 | |||
XM_011525980.2 | XP_011524282.1 | 54, 55 | 7263 | |||
XM_011525981.2 | XP_011524283.1 | 53, 54 | 7149 | |||
XM_017025743.1 | XP_016881232.1 | 37, 38 | 4965 | |||
XM_017025744.1 | XP_016881233.1 | 27, 28 | 3646 | |||
GenBank mRNA | AB107369.1 | - | 54, 55 | 7013 | ||
AB208853.1 | - | 9, 10 | 1239 | |||
AY327114.1 | - | 16, 17 | 2018 | |||
AY327115.1 | - | 54, 55 | 7013 | |||
AY327116.1 | - | 53, 54 | 6845 | |||
BC043618.1 | - | 11, 12 | 1591 | |||
L34155.1 | - | 17, 18 | 2153 | |||
X84900.1 | - | 19, 20 | 2432 | |||
X85107.1 | - | 17, 18 | 2204 | |||
X85108.1 | - | 19, 20 | 2432 |