Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000227.4 | NP_000218.3 | 19 | 2413 | ||
NM_001127717.2 | NP_001121189.2 | 55 | 7288 | |||
NM_001127718.2 | NP_001121190.2 | 18 | 2245 | |||
NM_198129.2 | NP_937762.2 | 56 | 7456 | |||
XM_011525978.2 | XP_011524280.1 | 56 | 7483 | |||
XM_011525979.2 | XP_011524281.1 | 56 | 7474 | |||
XM_011525980.2 | XP_011524282.1 | 56 | 7465 | |||
XM_011525981.2 | XP_011524283.1 | 55 | 7351 | |||
XM_011525982.2 | XP_011524284.1 | 54 | 7186 | |||
XM_017025743.1 | XP_016881232.1 | 39 | 5167 | |||
XM_017025744.1 | XP_016881233.1 | 29 | 3848 | |||
GenBank mRNA | AB107369.1 | - | 56 | 7215 | ||
AB208853.1 | - | 11 | 1441 | |||
AY327114.1 | - | 18 | 2220 | |||
AY327115.1 | - | 56 | 7215 | |||
AY327116.1 | - | 55 | 7047 | |||
BC043618.1 | - | 13 | 1793 | |||
L34155.1 | - | 19 | 2355 | |||
X84900.1 | - | 21 | 2634 | |||
X85107.1 | - | 19 | 2406 | |||
X85108.1 | - | 21 | 2634 |