Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005560.4 | NP_005551.3 | 70 | 9626 | ||
XM_006723796.3 | XP_006723859.1 | 70 | 9629 | |||
XM_011528818.2 | XP_011527120.1 | 69 | 9491 | |||
XM_011528819.2 | XP_011527121.1 | 68 | 9356 | |||
GenBank mRNA | AB011105.1 | - | 25 | 3411 | ||
AF443072.1 | - | 70 | 9626 | |||
AK074307.1 | - | 8 | 940 | |||
Z95636.1 | - | 10 | 1333 |