Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004139.4 | NP_004130.2 | 13 | 1392 | ||
GenBank mRNA | AF105067.1 | - | 13 | 1303 | ||
AK313625.1 | - | 13 | 1322 | |||
BC022256.1 | - | 13 | 1321 | 4767724 | ||
BQ028088.1 | - | 3 | 516 | |||
M35533.1 | - | 13 | 1274 |