Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF105067.1 | - | 6 | 630 | ||
AY344471.1 | - | 6 | 636 | |||
BC022256.1 | - | 6 | 648 | 4767724 |