Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270428.1 | NP_001257357.1 | 3 | 511 | ||
NM_002315.2 | NP_002306.1 | 3 | 843 | |||
NR_073006.1 | 3 | 843 | ||||
XM_006718228.3 | XP_006718291.1 | 3 | 1889 | |||
XM_011520098.2 | XP_011518400.1 | 3 | 378 | |||
XM_011520099.2 | XP_011518401.1 | 3 | 395 | |||
XM_011520100.1 | XP_011518402.1 | 2 | 242 | |||
GenBank mRNA | AW006226.1 | - | 2 | 469 | ||
BC039512.1 | - | 3 | 511 | |||
BC069673.1 | - | 3 | 385 | 7262648 | ||
BC069752.1 | - | 3 | 719 | 7262660 | ||
BC069793.1 | - | 3 | 719 | 7262577 | ||
BC096056.3 | - | 3 | 353 | 40000075 | ||
BC096057.3 | - | 3 | 352 | 40000076 | ||
BC096058.1 | - | 3 | 353 | 40000078 | ||
M26682.1 | - | 3 | 824 | |||
X64244.1 | - | 3 | 327 | |||
X64246.1 | - | 1 | 100 |