Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LMO7 SIRNA Assays ›
Gene Symbol:
LMO7
Gene Name:
LIM domain 7
Gene Aliases:
FBX20, FBXO20, LOMP
Chromosome Location:
Chr.13: 75620434 - 75859870 on Build GRCh38
UniGene ID:
Hs.207631
Species:
Human
siRNA ID
HSS106109
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LMO7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306080.1
NP_001293009.1
15
3215
NM_005358.5
NP_005349.3
14
3259
NM_015842.2
NP_056667.2
12
2906
XM_011535075.2
XP_011533377.1
15
2980
XM_011535078.1
XP_011533380.1
15
2980
XM_011535080.2
XP_011533382.1
15
2980
XM_011535082.1
XP_011533384.1
15
2980
XM_011535086.1
XP_011533388.1
14
2776
XM_011535094.1
XP_011533396.1
9
1603
XM_011535095.1
XP_011533397.1
12
1774
XM_017020574.1
XP_016876063.1
15
2980
XM_017020575.1
XP_016876064.1
15
2980
XM_017020576.1
XP_016876065.1
15
2980
XM_017020577.1
XP_016876066.1
15
2980
XM_017020578.1
XP_016876067.1
15
2980
XM_017020579.1
XP_016876068.1
15
2980
XM_017020580.1
XP_016876069.1
14
2776
XM_017020581.1
XP_016876070.1
14
2902
XM_017020582.1
XP_016876071.1
14
2776
XM_017020583.1
XP_016876072.1
14
2776
XM_017020584.1
XP_016876073.1
13
2698
XM_017020585.1
XP_016876074.1
13
2494
XM_017020586.1
XP_016876075.1
13
2494
XM_017020587.1
XP_016876076.1
13
2494
XM_017020588.1
XP_016876077.1
13
2494
XM_017020589.1
XP_016876078.1
13
2698
XM_017020590.1
XP_016876079.1
13
2494
XM_017020591.1
XP_016876080.1
14
3063
XM_017020592.1
XP_016876081.1
14
2946
XM_017020593.1
XP_016876082.1
13
2859
XM_017020594.1
XP_016876083.1
13
2742
XM_017020595.1
XP_016876084.1
13
2859
XM_017020596.1
XP_016876085.1
12
1774
XM_017020597.1
XP_016876086.1
11
1744
XM_017020598.1
XP_016876087.1
13
2859
XM_017020599.1
XP_016876088.1
12
2577
XM_017020600.1
XP_016876089.1
12
1774
XM_017020601.1
XP_016876090.1
12
2577
XM_017020602.1
XP_016876091.1
12
2460
XM_017020603.1
XP_016876092.1
12
2577
XM_017020604.1
XP_016876093.1
11
1857
GenBank mRNA
AB020665.2
-
12
2836
AF144237.1
-
3
719
AF330045.1
-
14
3258
AK055465.1
-
7
1871
AK092052.1
-
13
2178
AK304051.1
-
10
1716
FJ711162.1
-
15
2845
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
regulation of signaling
regulation of cell adhesion
ubiquitin-protein transferase activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0