Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000627.3 | NP_000618.3 | 12 | 1846 | ||
NM_001166264.1 | NP_001159736.1 | 12 | 1846 | |||
NM_001166265.1 | NP_001159737.1 | 12 | 1687 | |||
NM_001166266.1 | NP_001159738.1 | 12 | 1687 | |||
NM_206943.2 | NP_996826.2 | 16 | 2706 | |||
XM_005264317.3 | XP_005264374.1 | 16 | 2549 | |||
XM_005264318.3 | XP_005264375.1 | 16 | 2549 | |||
XM_011532853.2 | XP_011531155.1 | 16 | 2882 | |||
XM_011532855.2 | XP_011531157.1 | 16 | 2705 | |||
XM_011532856.2 | XP_011531158.1 | 16 | 2711 | |||
XM_011532857.2 | XP_011531159.1 | 16 | 2711 | |||
XM_011532859.2 | XP_011531161.1 | 16 | 2552 | |||
XM_011532860.2 | XP_011531162.1 | 15 | 2543 | |||
XM_011532861.2 | XP_011531163.1 | 16 | 2711 | |||
XM_011532862.2 | XP_011531164.1 | 16 | 2711 | |||
XM_017004108.1 | XP_016859597.1 | 16 | 2708 | |||
XM_017004109.1 | XP_016859598.1 | 16 | 2708 | |||
XM_017004110.1 | XP_016859599.1 | 16 | 2708 | |||
GenBank mRNA | AB208801.1 | - | 12 | 1645 | ||
AF489528.1 | - | 12 | 1702 | |||
AK298027.1 | - | 6 | 415 | |||
AK299912.1 | - | 7 | 644 | |||
AL703596.1 | - | 6 | 537 | |||
BC130289.1 | - | 12 | 1786 | 40146320 | ||
BC144127.1 | - | 12 | 1629 | |||
BC144128.1 | - | 12 | 1786 | 9052648 | ||
M34057.1 | - | 12 | 1792 |