Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111097.2 | NP_001104567.1 | 6 | 668 | ||
NM_002350.3 | NP_002341.1 | 6 | 731 | |||
XM_011517529.2 | XP_011515831.1 | 5 | 3116 | |||
XM_017013415.1 | XP_016868904.1 | 5 | 3116 | |||
XM_017013416.1 | XP_016868905.1 | 6 | 407 | |||
GenBank mRNA | AK290494.1 | - | 6 | 641 | ||
AK298677.1 | - | 4 | 319 | |||
BC059394.1 | - | 6 | 660 | |||
BC068551.1 | - | 6 | 660 | |||
BC075001.2 | - | 5 | 474 | 30915399 | ||
BC075002.2 | - | 5 | 474 | 30915611 | ||
BC126456.1 | - | 7 | 568 | 8992172 | ||
BC126458.1 | - | 7 | 615 | 8992174 | ||
CB990993.1 | - | 6 | 693 | |||
M16038.1 | - | 6 | 746 | |||
M79321.1 | - | 5 | 410 |