Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114937.2 | NP_001108409.1 | 3 | 672 | ||
NM_002351.4 | NP_002342.1 | 3 | 672 | |||
GenBank mRNA | AB586694.1 | - | 3 | 413 | ||
AF072930.1 | - | 3 | 397 | |||
AF073019.1 | - | 3 | 397 | |||
AF100539.1 | - | 3 | 310 | |||
AF100540.1 | - | 2 | 301 | |||
AF100541.1 | - | 3 | 365 | |||
AF100542.1 | - | 3 | 288 | |||
AF100543.1 | - | 2 | 246 | |||
AK311645.1 | - | 2 | 171 | |||
AK311911.1 | - | 3 | 346 | |||
AL023657.1 | - | 3 | 610 | |||
BC020732.1 | - | 3 | 404 | 4702653 | ||
CR542031.1 | - | 3 | 311 | |||
CR542043.1 | - | 3 | 311 |