Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003652.3 | NP_001003652.1 | 4 | 779 | ||
NM_001135937.2 | NP_001129409.1 | 3 | 689 | |||
NM_005901.5 | NP_005892.1 | 4 | 671 | |||
XM_005258259.3 | XP_005258316.1 | 5 | 912 | |||
XM_006722451.3 | XP_006722514.1 | 5 | 924 | |||
XM_011525984.2 | XP_011524286.1 | 4 | 268 | |||
XM_011525985.2 | XP_011524287.1 | 6 | 971 | |||
XM_017025745.1 | XP_016881234.1 | 6 | 1010 | |||
XM_017025746.1 | XP_016881235.1 | 4 | 822 | |||
XM_017025747.1 | XP_016881236.1 | 5 | 897 | |||
XM_017025748.1 | XP_016881237.1 | 6 | 983 | |||
XM_017025749.1 | XP_016881238.1 | 4 | 606 | |||
XM_017025750.1 | XP_016881239.1 | 4 | 826 | |||
GenBank mRNA | AF027964.1 | - | 4 | 485 | ||
AK299218.1 | - | 3 | 385 | |||
AK312963.1 | - | 3 | 508 | |||
BC014840.1 | - | 4 | 627 | 4449540 | ||
BC025699.1 | - | 4 | 485 | 5221801 | ||
BT007422.1 | - | 3 | 379 | |||
DC371139.1 | - | 3 | 385 | |||
U59911.1 | - | 4 | 478 | |||
U65019.1 | - | 3 | 415 | |||
U68018.1 | - | 3 | 434 |