Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005359.5 | NP_005350.1 | 9 | 1582 | ||
GenBank mRNA | AK290770.1 | - | 9 | 1244 | ||
BC002379.2 | - | 10 | 1277 | 2961238 | ||
BX647129.1 | - | 5 | 3046 | |||
KU178168.1 | - | 8 | 1044 | |||
KU178169.1 | - | 8 | 1044 | |||
U44378.1 | - | 8 | 1172 |