Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001419.2 | NP_001001419.1 | 4 | 772 | ||
NM_001001420.2 | NP_001001420.1 | 2 | 613 | |||
NM_005903.6 | NP_005894.3 | 3 | 688 | |||
XM_017009470.1 | XP_016864959.1 | 4 | 904 | |||
GenBank mRNA | AF009678.1 | - | 1 | 340 | ||
AF010601.1 | - | 1 | 457 | |||
AK313397.1 | - | 3 | 688 | |||
BC009682.2 | - | 1 | 494 | 3906006 | ||
BX510058.1 | - | 2 | 614 | |||
BX648328.1 | - | 3 | 627 | |||
CR457424.1 | - | 1 | 326 | |||
CR749366.1 | - | 3 | 627 | |||
CR749473.1 | - | 3 | 615 | |||
U59913.1 | - | 1 | 456 | |||
U73825.1 | - | 1 | 475 |