Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190821.1 | NP_001177750.1 | 3, 4 | 1028 | ||
NM_001190822.1 | NP_001177751.1 | 3, 4 | 236 | |||
NM_001190823.1 | NP_001177752.1 | 1, 2 | 329 | |||
NM_005904.3 | NP_005895.1 | 3, 4 | 1031 | |||
GenBank mRNA | AF010193.1 | - | 3, 4 | 1039 | ||
AF015261.1 | - | 3, 4 | 744 | |||
AK131394.1 | - | 1, 2 | 271 | |||
AK301535.1 | - | 3, 4 | 236 | |||
BC074818.2 | - | 3, 4 | 778 | 30915321 | ||
BC074819.2 | - | 3, 4 | 778 | 30915535 | ||
BP363832.1 | - | 1, 2 | 329 | |||
DA882147.1 | - | 3, 4 | 460 |