Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN2C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256494.1 | NP_001243423.1 | 14, 15 | 1757 | ||
NM_001256495.1 | NP_001243424.1 | 14, 15 | 1757 | |||
NM_001256496.1 | NP_001243425.1 | 12, 13 | 1460 | |||
NM_006715.3 | NP_006706.2 | 14, 15 | 1757 | |||
XM_005254384.2 | XP_005254441.1 | 14, 15 | 1810 | |||
XM_017022186.1 | XP_016877675.1 | 13, 14 | 2105 | |||
XM_017022187.1 | XP_016877676.1 | 13, 14 | 2105 | |||
XM_017022188.1 | XP_016877677.1 | 12, 13 | 1513 | |||
GenBank mRNA | AB209141.1 | - | 4128 | |||
AF044414.2 | - | 14, 15 | 1769 | |||
AK225145.1 | - | 12, 13 | 1437 | |||
AK311065.1 | - | 13, 14 | 1609 | |||
AL136876.1 | - | 14, 15 | 1718 | |||
BC050550.1 | - | 14, 15 | 1724 | 5529534 | ||
BC080191.1 | - | 14, 15 | 1722 | 6149811 |