Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN2C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256494.1 | NP_001243423.1 | 4 | 414 | ||
NM_001256495.1 | NP_001243424.1 | 4 | 414 | |||
NM_001256496.1 | NP_001243425.1 | 4 | 414 | |||
NM_006715.3 | NP_006706.2 | 4 | 414 | |||
XM_005254384.2 | XP_005254441.1 | 4 | 467 | |||
XM_017022186.1 | XP_016877675.1 | 3 | 762 | |||
XM_017022187.1 | XP_016877676.1 | 3 | 762 | |||
XM_017022188.1 | XP_016877677.1 | 4 | 467 | |||
GenBank mRNA | AB209141.1 | - | 292 | |||
AF044414.2 | - | 4 | 426 | |||
AK225145.1 | - | 4 | 391 | |||
AK294892.1 | - | 4 | 393 | |||
AK295265.1 | - | 4 | 723 | |||
AK311065.1 | - | 3 | 266 | |||
AL136876.1 | - | 4 | 375 | |||
BC050550.1 | - | 4 | 381 | 5529534 | ||
BC080191.1 | - | 4 | 379 | 6149811 | ||
DC342919.1 | - | 4 | 417 |