Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286124.1 | NP_001273053.1 | 18 | 2932 | ||
NM_001286126.1 | NP_001273055.1 | 17 | 2884 | |||
NM_001286128.1 | NP_001273057.1 | 16 | 2818 | |||
NM_018650.4 | NP_061120.3 | 18 | 2929 | |||
XM_005273134.4 | XP_005273191.1 | 17 | 2754 | |||
XM_006711326.3 | XP_006711389.1 | 18 | 2193 | |||
XM_011509561.2 | XP_011507863.1 | 18 | 2928 | |||
XM_011509562.2 | XP_011507864.1 | 18 | 2545 | |||
XM_017001305.1 | XP_016856794.1 | 19 | 2389 | |||
XM_017001306.1 | XP_016856795.1 | 18 | 2426 | |||
XM_017001307.1 | XP_016856796.1 | 18 | 2781 | |||
GenBank mRNA | AB040910.1 | - | 16 | 2775 | ||
AF154845.1 | - | 18 | 2555 | |||
AK295506.1 | - | 17 | 2382 | |||
AK314614.1 | - | 18 | 2592 | |||
AL512724.1 | - | 1 | 276 | |||
BC113869.1 | - | 18 | 2465 | 40036731 | ||
BC114478.1 | - | 18 | 2465 | 40036730 | ||
KJ534879.1 | - | 17 | 2247 | |||
KJ534880.1 | - | 12 | 1689 | |||
KJ534881.1 | - | 17 | 2244 | |||
KJ535089.1 | - | 18 | 2288 |