Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042539.2 | NP_001036004.1 | 3 | 1252 | ||
NM_001276275.1 | NP_001263204.1 | 4 | 1067 | |||
NM_002383.3 | NP_002374.2 | 3 | 1252 | |||
NR_074080.1 | 2 | 401 | ||||
XM_006721047.3 | XP_006721110.1 | 4 | 1039 | |||
XM_006721048.3 | XP_006721111.1 | 3 | 1412 | |||
GenBank mRNA | AB209061.1 | - | 1 | 784 | ||
BC041629.1 | - | 2 | 402 | 4309842 | ||
BI198991.1 | - | 2 | 286 | |||
D85131.1 | - | 4 | 1253 | |||
EF059746.1 | - | 3 | 1087 | |||
FJ532357.1 | - | 4 | 1057 | |||
M93339.1 | - | 3 | 1138 | |||
M94046.1 | - | 3 | 1109 | |||
U33819.1 | - | 3 | 1267 |