Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085377.1 | NP_001078846.1 | 9 | 1889 | ||
NM_002387.2 | NP_002378.1 | 7 | 1433 | |||
XM_005271991.2 | XP_005272048.1 | 7 | 1467 | |||
XM_017009473.1 | XP_016864962.1 | 9 | 1488 | |||
XM_017009474.1 | XP_016864963.1 | 7 | 4153 | |||
GenBank mRNA | AK096212.1 | - | 6 | 1035 | ||
AK128596.1 | - | 9 | 1901 | |||
AK300259.1 | - | 7 | 1456 | |||
BC009279.2 | - | 7 | 1433 | 4111706 | ||
BC018919.2 | - | 7 | 1433 | 4131685 | ||
BX537952.1 | - | 7 | 904 | |||
BX647710.1 | - | 9 | 1363 | |||
M62397.1 | - | 7 | 1123 |